Learn More
Klebsiella Monoclonal Antibody (73/28), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA183282
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody has detected Klebsiella oxytoca and other serotypes in fresh isolates, including those fixed by phenol, formalin or boiling. Mouse anti Klebsiella SPP antibody, clone 73/28 is reactive with Klebsiella spp. serotypes.
Klebsiella Monoclonal antibody specifically detects Klebsiella in Bacteria samples. It is validated for ELISA, Immunocytochemistry.Spezifikation
Klebsiella | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Native K. aerogenes preparation | |
200 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry | |
73/28 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Bacteria | |
Antibody | |
IgG2a |