Learn More
KLF15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA124490
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-24490 detects KLF15 in Human samples. PA1-24490 has been successfully used in Immunohistochemistry and Western Blot procedures. PA1-24490 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human KLF15. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
KLF15 is a Cys2-His2 zinc finger gene. It is found abundantly expressed in the liver, kidneys, heart, and skeletal muscle.Spezifikation
KLF15 | |
Polyclonal | |
Unconjugated | |
Q9UIH9 | |
KLF15 | |
Synthetic peptide region of human KLF15. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
KLF15 | |
DKFZp779M1320, KKLF | |
Rabbit | |
Protein A | |
RUO | |
28999 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |