Learn More
KLF2 Monoclonal Antibody (665333), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524300
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) KLF17 and no cross-reactivity with rhKLF1, 4, 5, 6, 10, 12, 17, recombinant mouse KLF4 or 15 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
KLF2 Monoclonal antibody specifically detects KLF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Spezifikation
KLF2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9Y5W3 | |
KLF2 | |
E. coli-derived recombinant human KLF2 Pro71-Pro168 (predicted) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry | |
665333 | |
Unconjugated | |
KLF2 | |
KLF2; Krueppel-like factor 2; Kruppel like factor 2; Kruppel-like factor 2; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; Lklf; LOW QUALITY PROTEIN: Krueppel-like factor 2; lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor | |
Mouse | |
Protein A/G | |
RUO | |
10365 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |