missing translation for 'onlineSavingsMsg'
Learn More

KLF4 Recombinant Polyclonal Antibody (1HCLC), Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Marke:  Invitrogen 710659

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15450664

  • 469.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody is predicted to react with Monkey, Pig and Rabbit. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

KLF4 Recombinant Polyclonal antibody specifically detects KLF4 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

KLF4
Recombinant Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O43474
KLF4
full-length recombinant protein corresponding to human KLF4 (aa 1-118)
100 μg
Primary
Human
Antibody
IgG
Immunocytochemistry, Western Blot
1HCLC
Unconjugated
KLF4
endothelial Kruppel-like zinc finger protein; epithelial zinc finger protein EZF; Ezf; Gklf; gut Kruppel-like factor; gut-enriched krueppel-like factor; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; KLF4; Krueppel-like factor 4; Kruppel like factor 4; Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); PNAS-24; PNAS-25; RP11-150J11.1; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; Zie
Rabbit
Protein A
RUO
9314
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote