Learn More
KLF4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527441
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa. Predicted reactivity: Mouse (89%), Rat (89%), Cat (93%), Pig (93%), Sheep (92%), Rhesus Monkey (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KLF4 Polyclonal antibody specifically detects KLF4 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KLF4 | |
Polyclonal | |
Unconjugated | |
KLF4 | |
endothelial Kruppel-like zinc finger protein; epithelial zinc finger protein EZF; Ezf; Gklf; gut Kruppel-like factor; gut-enriched krueppel-like factor; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; KLF4; Krueppel-like factor 4; Kruppel like factor 4; Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); PNAS-24; PNAS-25; RP11-150J11.1; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; Zie | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16600, 9314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43474, Q60793 | |
KLF4 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 212 of Human KLF4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |