Learn More
KLHDC8A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528806
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLaS3, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KLHDC8A Polyclonal antibody specifically detects KLHDC8A in Human samples. It is validated for Western Blot.Spezifikation
KLHDC8A | |
Polyclonal | |
Unconjugated | |
KLHDC8A | |
A630065K24Rik; kelch domain containing 8A; kelch domain-containing protein 8A; KLHDC8A; RGD1305132; S-delta-E1; substitute for delta-EGFR expression 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55220 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8IYD2 | |
KLHDC8A | |
Synthetic peptide corresponding to a region within amino acids 287 and 350 of Human KLHDC8A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |