Learn More
KLHL21 Monoclonal Antibody (2H4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515654
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15654 targets KLHL21 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15654 immunogen is purified recombinant fragment of human KLHL21 expressed in E. Coli.<br>. MA5-15654 detects KLHL21 which has a predicted molecular weight of approximately 66kDa.
KLHL21 Monoclonal antibody specifically detects KLHL21 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
KLHL21 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q9UJP4 | |
KLHL21 | |
Purified recombinant fragment of human KLHL21 expressed in E. Coli. | |
RUO | |
9903 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
2H4 | |
Unconjugated | |
KLHL21 | |
1810045K06Rik; D330008A20; kelch like family member 21; kelch-like 21; kelch-like family member 21; kelch-like protein 21; KIAA0469; Klhl21; MGC99635; mKIAA0469; RGD1305863 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |