Learn More
KLHL22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532054
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, HeLa, HepG2. Predicted reactivity: Mouse (93%), Rat (93%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KLHL22 Polyclonal antibody specifically detects KLHL22 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KLHL22 | |
Polyclonal | |
Unconjugated | |
KLHL22 | |
2610318I18Rik; kelch like family member 22; KELCHL; kelch-like 22; kelch-like family member 22; kelch-like protein 22; KLHL22; RGD1306288 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84861 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.04 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q53GT1 | |
KLHL22 | |
Recombinant fragment corresponding to a region within amino acids 412 and 634 of Human KLHL22 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |