missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL24 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525162
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse and rat based on sequence homology.
KLHL24 Polyclonal antibody specifically detects KLHL24 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KLHL24 | |
Polyclonal | |
Unconjugated | |
KLHL24 | |
1110046J11Rik; 4930429H24Rik; 6530402O20Rik; C85082; DRE1; kainate receptor interacting protein for GluR6; kainate receptor-interacting protein for GluR6; kelch like family member 24; kelch-like 24; kelch-like family member 24; kelch-like protein 24; Klhl24; KRIP6; Protein DRE1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
54800 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q6TFL4 | |
KLHL24 | |
KLH conjugated synthetic peptide between 182-211 amino acids from the central region of human KLHL24 | |
400 μL | |
Primary | |
Human | |
Antibody | |
Ig |