Learn More
ENC-2 Monoclonal Antibody (1B7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515656
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15656 targets KLHL25 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15656 immunogen is purified recombinant fragment of human KLHL25 expressed in E. Coli. <br>. MA5-15656 detects KLHL25 which has a predicted molecular weight of approximately 65kDa.
ENC-2 Monoclonal antibody specifically detects ENC-2 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
ENC-2 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q9H0H3 | |
KLHL25 | |
Purified recombinant fragment of human KLHL25 expressed in E. Coli. | |
RUO | |
64410 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1B7 | |
Unconjugated | |
KLHL25 | |
2810402K13Rik; BC027373; BTB/POZ KELCH domain protein; ectodermal-neural cortex 2; ectoderm-neural cortex protein 2; ectoderm-neural cortex-1; Enc-1; ENC2; ENC-2; FLJ12587; FLJ45015; kelch like family member 25; kelch-like 25; kelch-like family member 25; kelch-like protein 25; KLHL25; RGD1310815 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |