Learn More
KLHL26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535910
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Human (99%), Mouse (84%), Rat (84%), Zebrafish (80%), Rhesus Monkey (99%).
KLHL26 Polyclonal antibody specifically detects KLHL26 in Human samples. It is validated for Immunocytochemistry.Spezifikation
KLHL26 | |
Polyclonal | |
Unconjugated | |
KLHL26 | |
C630013N10Rik; kelch like family member 26; kelch-like 26; kelch-like family member 26; kelch-like protein 26; KLHL26; Klkl26; RGD1312034 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q53HC5 | |
KLHL26 | |
Recombinant fragment corresponding to a region within amino acids 293 and 603 of KLHL26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |