missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klotho Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518447
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse and rat based on sequence homology.
Klotho Polyclonal antibody specifically detects Klotho in Human samples. It is validated for Immunohistochemistry.Spezifikation
Klotho | |
Polyclonal | |
Unconjugated | |
KL | |
alpha-kl; HFTC3; Kl; Klotho; Klotho peptide; secreted form of Klotho protein | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9365 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UEF7 | |
KL | |
Synthetic peptide sequence (KRDDAKYMYYLKK) corresponding to the internal amino acids of KL | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |