missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKG2E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542708
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MSKQRGTFSE VSLAQDPKWQ QRKPKGNKSS ISGTEQEIFQ VELNLQNASL Sequence homology: Human: 100%; Mouse: 79%
NKG2E Polyclonal antibody specifically detects NKG2E in Human samples. It is validated for Western Blot.Spezifikation
NKG2E | |
Polyclonal | |
Unconjugated | |
KLRC3 | |
killer cell lectin like receptor C3; killer cell lectin-like receptor subfamily C, member 2; killer cell lectin-like receptor subfamily C, member 3; Klrc2; KLRC3; natural killer cell group 2E cell receptor; NK cell receptor E; NKG2E; NKG2-E; NKG2-E type II integral membrane protein; NKG2-E-activating NK receptor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3823 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q07444 | |
KLRC3 | |
Synthetic peptide directed towards the N-terminal of human KLRC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |