missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMO Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535353
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
KMO Polyclonal antibody specifically detects KMO in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KMO | |
Polyclonal | |
Unconjugated | |
KMO | |
AI046660; dJ317G22.1; HAMAP-Rule:MF_03018}; KMO; kmo {ECO:0000312; kynurenine 3-hydroxylase; kynurenine 3-hydroxylase {ECO:0000255; Kynurenine 3-monooxygenase; kynurenine 3-monooxygenase (kynurenine 3-hydroxylase); kynurenine 3-monooxygenase {ECO:0000255; RGD:620610} | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8564 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide | |
O15229 | |
KMO | |
KLH conjugated synthetic peptide between 155-182 amino acids from the central region of human KMO | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |