missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHKA1/PHKA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538611
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Phosphorylase b kinase catalyzes the phosphorylation of serine in certain substrates, including troponin I. The alpha chain may bind calmodulin.Spezifikation
PHKA1/PHKA2 | |
Polyclonal | |
Unconjugated | |
PHKA1 | |
PHKA1, PHKA, phosphorylase b kinase regulatory subunit alpha skeletal muscle isoform, phosphorylase b kinase regulatory subunit alpha, skeletal muscle isoform, phosphorylase kinase alpha M subunit, Phosphorylase b kinase regulatory subunit alpha, liver isoform, Phosphorylase kinase alpha L subunit, PHKA2, PHKLA, PYK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110094, 18679, 5255, 5256, 64561, 678739 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P46019, P46020, P18826, Q8BWJ3, Q64649 | |
PHKA1, PHKA2 | |
A synthetic peptide derived from the internal region of human KPB1/2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |