missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534372
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-34372 can be used with blocking peptide PEP-1415.
KPNA7 Polyclonal antibody specifically detects KPNA7 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KPNA7 | |
Polyclonal | |
Unconjugated | |
KPNA7 | |
importin alpha 8; importin subunit alpha-8; IPOA8; karyopherin 7; karyopherin alpha 7 (importin alpha 8); karyopherin subunit alpha 7; Karyopherin subunit alpha-7; KPNA7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
402569 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
A9QM74 | |
KPNA7 | |
A 13 amino acid synthetic peptide near the amino terminus of human KPNA7 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |