Learn More
KSHV ORF8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519852
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-19852 targets KSHV ORF8 in indirect ELISA, WB applications and shows reactivity with Human samples. The PA5-19852 immunogen is synthetic peptide, corresponding to amino acids sequence of KSHV ORF8. <br>.
KSHV ORF8 Polyclonal antibody specifically detects KSHV ORF8 in Virus samples. It is validated for ELISA, Western Blot.Spezifikation
KSHV ORF8 | |
Polyclonal | |
Unconjugated | |
Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus | |
Synthetic peptide, corresponding to amino acids sequence of KSHV ORF8. | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA, Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
Rabbit | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |