missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514027
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
KSR1 Polyclonal antibody specifically detects KSR1 in Human samples. It is validated for Western Blot.Spezifikation
KSR1 | |
Polyclonal | |
Unconjugated | |
KSR1 | |
AW492498; B-KSR1; CAP kinase; D11Bhm183e; D11Bhm184e; hb; HB protein; kinase KSR1; Kinase suppressor of Ras 1; Ksr; KSR1; mKSR1; OTTHUMP00000162931; protein Hb; RSU2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
8844 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q8IVT5 | |
KSR1 | |
KLH conjugated synthetic peptide between 159-194 amino acids from the N-terminal region of human KSR | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |