Learn More
KCNK4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541045
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LVSGALVFRA LEQPHEQQAQ RELGEVREKF LRAHPCVSDQ ELGLLIKEVA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Pig: 100%; Rat: 85%
KCNK4 Polyclonal antibody specifically detects KCNK4 in Human samples. It is validated for Western Blot.Spezifikation
KCNK4 | |
Polyclonal | |
Unconjugated | |
KCNK4 | |
K2p4.1; K2P4.1 potassium channel; Kcnk4; KT4.1; KT4.1b; LOW QUALITY PROTEIN: potassium channel subfamily K member 4; MLZ-622; Potassium channel subfamily K member 4; potassium channel, subfamily K, member 4; potassium channel, two pore domain subfamily K, member 4; potassium inwardly-rectifying channel subfamily K member 4; potassium inwardly-rectifying channel, subfamily K, member 4; potassium two pore domain channel subfamily K member 4; protein expressed in male leptotene and zygotene spermatocytes 622; Tex40; TRAAK; TRAAK1; TRAAKt; TWIK-related arachidonic acid-stimulated potassium channel protein; Two pore K(+) channel KT4.1; two pore K+ channel KT4.1; two pore potassium channel KT4.1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
50801 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NYG8 | |
KCNK4 | |
Synthetic peptide directed towards the N-terminal of human KCNK4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |