missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KTI12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545752
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VPKELEREES GAAESPALVT PDSEKSAKHG SGAFYSPELL EALTLRFEAP Sequence homology: Human: 100%
KTI12 Polyclonal antibody specifically detects KTI12 in Human samples. It is validated for Western Blot.Spezifikation
KTI12 | |
Polyclonal | |
Unconjugated | |
KTI12 | |
KTI12; KTI12 chromatin associated homolog; KTI12 homolog, chromatin associated; KTI12 homolog, chromatin associated (S. cerevisiae); protein KTI12 homolog; RP11-91A18.3; SBBI81; TOT4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
112970 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96EK9 | |
KTI12 | |
Synthetic peptide directed towards the middle region of human KTI12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |