missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ku70 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525915
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Ku70 Polyclonal antibody specifically detects Ku70 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Ku70 | |
Polyclonal | |
Unconjugated | |
XRCC6 | |
5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; 70kDa; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CT; CTA-216E10.7; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; CTCBF; DNA repair protein XRCC6; G22p1; Ku autoantigen p70 subunit; ku autoantigen protein p70 homolog; Ku autoantigen, 70kDa; Ku p70; Ku70; Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa); Kup70; Lupus Ku autoantigen protein p70; ML8; OTTHUMP00000028581; thyroid autoantigen; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; XELAEV_18023498mg; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair complementing defective repair in Chinese hamste | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
14375, 2547 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P12956, P23475 | |
XRCC6 | |
KLH conjugated synthetic peptide between 432-461 amino acids from the central region of human Ku70 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |