missing translation for 'onlineSavingsMsg'
Learn More

KV1.6 (KCNA6) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA519498

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11357463

  • 514.00 € / 100 Mikrogramm
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody is predicted to react with human based on sequence homology.

KV1.6 (KCNA6) Polyclonal antibody specifically detects KV1.6 (KCNA6) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

KV1.6 (KCNA6)
Polyclonal
Unconjugated
KCNA6
FLJ25134; HBK2; human brain potassium channel-2; KCNA6; KV1.6; kv2; MK1.6; potassium channel, voltage gated shaker related subfamily A, member 6; potassium channel-Kv2; potassium voltage gated channel, shaker related subfamily, member 6; potassium voltage gated channel, shaker related, subfamily, member 6; potassium voltage-gated channel subfamily A member 6; potassium voltage-gated channel, shaker-related subfamily, member 6; potassium voltage-gated channel, shaker-related, subfamily, member 6; PPP1R96; protein phosphatase 1, regulatory subunit 96; RCK2; voltage-gated potassium channel HBK2; voltage-gated potassium channel protein Kv1.6; voltage-gated potassium channel subunit Kv1.6; voltage-gated potassium channel subunit Kv2
Rabbit
Antigen affinity chromatography
RUO
16494, 3742, 64358
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
1.0 mg/mL
PBS with 0.02% sodium azide; pH 7.4
P17658, P17659, Q61923
KCNA6
Synthetic peptide conjugated to KLH derived from within residues 500 to the C-terminus of Human Kv1.6
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote