missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV3.3 (KCNC3) Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA537871
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This antibody is predicted to react with human and pig based on sequence homology.
KV3.3 (KCNC3) Polyclonal antibody specifically detects KV3.3 (KCNC3) in Mouse, Rat samples. It is validated for Western Blot.Spezifikation
KV3.3 (KCNC3) | |
Polyclonal | |
Unconjugated | |
KCNC3 | |
KCNC3; Kcr2-3; KShIIID; KV3.3; LOW QUALITY PROTEIN: potassium voltage-gated channel subfamily C member 3; potassium channel, voltage gated Shaw related subfamily C, member 3; potassium channel, voltage gated Shaw-related subfamily C, member 3; potassium voltage gated channel, Shaw-related subfamily, member 3; potassium voltage-gated channel subfamily C member 3; potassium voltage-gated channel, Shaw-related subfamily, member 3; SCA13; Shaw type potassium channel Kv3.3; Shaw-related voltage-gated potassium channel protein 3; voltage-gated potassium channel protein KV3.3; voltage-gated potassium channel subunit Kv3.3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
117101, 16504 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q01956, Q63959 | |
KCNC3 | |
Peptide with sequence C-HISNKTVTQASP, from the internal region of KCNC3 (aa317-328) | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |