Learn More
KV4.3 (KCND3) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530942
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: IMR32, U87-MG. Predicted reactivity: Mouse (98%), Rat (99%), Rabbit (97%), Chicken (89%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KV4.3 (KCND3) Polyclonal antibody specifically detects KV4.3 (KCND3) in Human samples. It is validated for Western Blot.Spezifikation
KV4.3 (KCND3) | |
Polyclonal | |
Unconjugated | |
KCND3 | |
AW045978; BRGDA9; KCND3; KCND3L; KCND3S; Kncd3; KSHIVB; KV4.3; potassium channel alpha subunit Kv4.3 short form; potassium channel Kv4.3; potassium channel Kv4.3L; potassium channel Kv4.3M; potassium channel, voltage gated Shal related subfamily D, member 3; potassium channel, voltage-gated Shal-related subfamily D, member 3; potassium ionic channel Kv4.3; potassium voltage gated channel, Shal-related family, member 3; potassium voltage-gated channel long; potassium voltage-gated channel subfamily D member 3; potassium voltage-gated channel, Shal-related family, member 3; potassium voltage-gated channel, Shal-related subfamily, member 3; SCA19; SCA22; sha1-related potassium channel Kv4.3; voltage-gated K+ channel; voltage-gated potassium channel subunit Kv4.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3752 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.88 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UK17 | |
KCND3 | |
Recombinant fragment corresponding to a region within amino acids 462 and 655 of Human Kv4.3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |