missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517059
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
SSB Polyclonal antibody specifically detects SSB in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
SSB | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P05455, P32067, P38656 | |
SSB | |
Synthetic peptide corresponding to the sequence of human La Antigen | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
SSB | |
autoantigen La; La; la autoantigen; La autoantigen homolog; La protein; la ribonucleoprotein; La ribonucleoprotein domain family, member 3; La/SSB; LARP3; lupus La antigen; lupus La protein; Lupus La protein homolog; Lupus LA protein homolog (LA ribonucleoprotein) (LA autoantigen homolog); lupus La protein-like; mRNA for autoantigen; OTTHUMP00000204777; sjoegren syndrome type B antigen; Sjogren syndrome antigen B; Sjogren syndrome antigen B (autoantigen La); small RNA binding exonuclease protection factor La; Ssb; SS-B; SS-B/La protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20823, 6741, 81783 | |
-20°C | |
Liquid |