missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactoferrin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA131254
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-31254 detects Lactoferrin from human samples.
Lactoferrin Polyclonal antibody specifically detects Lactoferrin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Lactoferrin | |
Polyclonal | |
Unconjugated | |
LTF | |
Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; LTF; minisatellite 10r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4057 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.80 mg/mL | |
tris glycine with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.9 | |
P02788 | |
LTF | |
purified full length human native protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |