Learn More
Lambda Light Chain Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA516651
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-16651 targets Lambda Light Chain in IHC (P) applications and shows reactivity with Human, mouse, Non-human primate, and Rat samples. The PA5-16651 immunogen is lambda light chains purified from human Bence Jones proteins.
Lambda Light Chain Polyclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Lambda Light Chain | |
Polyclonal | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P01701, P0CG04 | |
IGL, IGLC1, IGLV, IGLV1-51 | |
Lambda light chains purified from human Bence Jones proteins | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IGLC1 | |
Bence Jones Protein; BJP; Clambda1; Ig lambda chain V-I region BL2; IgL; Igl@; IGLC; IGLC 1/2/3; IGLC1; Igl-C1; IGLL5; IGLV; IGLV@; immmunoglobulin lambda light chain variable region; Immunoglobulin L; immunoglobulin lambda chain complex; immunoglobulin lambda constant 1; immunoglobulin lambda constant 7; immunoglobulin lambda light chain; immunoglobulin lambda light chain constant region; immunoglobulin lambda light chain V-J region; immunoglobulin lambda variable cluster; Lambda LC; lambda-immunoglobulin; LOC100060608; Mcg Marker; Paraprotein | |
Rabbit | |
purified | |
RUO | |
28820, 3535, 3537, 3546 | |
4° C | |
Liquid |