Learn More
Lamin B2 Monoclonal Antibody (LN43), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA106104
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-06104 detects lamin B2 in human, mouse, porcine, Xenopus laevis, zebrafish and hamster samples. MA1-06104 has sucessfully been used in FACS, immunohistochemistry, and Western blotting procedures. The MA1-06104 immunogen is a detergent insoluble fraction of potoroo cell line PtK1. Store at 4oC, or in small aliquots at -20oC.
Lamin B2 Monoclonal antibody specifically detects Lamin B2 in Hamster, Human, Mouse, Porcine, X. laevis, Zebrafish samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
Lamin B2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P21619, Q03252 | |
LMNB2, lmnb2.S | |
Detergent insoluble fraction of potoroo cell line PtK1. | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Porcine, X. laevis, Zebrafish | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
LN43 | |
Unconjugated | |
LMNB2 | |
EPM9; fc15d06; im:7142331; LAMB2; lamin; lamin A/C; lamin B2; lamin B2 S homeolog; lamin B3; lamin-B2; Lamin-L(II); LMN2; LMNA; lmnb2; lmnb2.S; lmnb2-a; lmnb2-b; MGC2721; RGD1563803; unnamed protein product; wu:fb94e05; wu:fb95e12; wu:fc15d06; wu:fc49h03; XELAEV_18009631mg | |
Mouse | |
purified | |
RUO | |
100620983, 100766421, 16907, 30196, 394291, 84823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |