missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin B2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529121
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: U87-MG. Predicted reactivity: Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Lamin B2 Polyclonal antibody specifically detects Lamin B2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Lamin B2 | |
Polyclonal | |
Unconjugated | |
LMNB2 | |
EPM9; fc15d06; im:7142331; LAMB2; lamin; lamin A/C; lamin B2; lamin B2 S homeolog; lamin B3; lamin-B2; Lamin-L(II); LMN2; LMNA; lmnb2; lmnb2.S; lmnb2-a; lmnb2-b; MGC2721; RGD1563803; unnamed protein product; wu:fb94e05; wu:fb95e12; wu:fc15d06; wu:fc49h03; XELAEV_18009631mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q03252 | |
LMNB2 | |
Recombinant protein fragment corresponding to a region within amino acids 277 and 560 of Human Lamin B2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |