Learn More
Laminin beta-3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529161
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431. Predicted reactivity: Mouse (87%), Rat (87%), Rabbit (83%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Laminin beta-3 Polyclonal antibody specifically detects Laminin beta-3 in Human samples. It is validated for Western Blot.Spezifikation
Laminin beta-3 | |
Polyclonal | |
Unconjugated | |
LAMB3 | |
AI1A; BM600-125KDA; Epiligrin subunit bata; kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; LAM5; Lamb3; Laminin B1k chain; laminin beta 3; laminin chain; laminin S B3 chain; laminin subunit beta 3; laminin subunit beta-3; laminin, beta 3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); laminin-5 subunit beta; LAMNB1; Nicein subunit beta; nicein, 125kD; nicein, 125kDa; nicein-125k; nicein-125kDa; RP1-272L16.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13751 | |
LAMB3 | |
Recombinant fragment corresponding to a region within amino acids 749 and 1017 of Human Laminin beta 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |