missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP2 Monoclonal Antibody (H4B4), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen A15464
Weitere Details : Netto-Gewicht : 0.00280kg
Beschreibung
LAMP2 Monoclonal antibody specifically detects LAMP2 in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry.Spezifikation
LAMP2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P13473 | |
LAMP2 | |
Human CD107b from Human adherent peripheral blood cells | |
100 μg | |
Primary | |
Human, Non-human Primate, Rhesus Monkey | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
H4B4 | |
Alexa Fluor 647 | |
LAMP2 | |
CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2 | |
Mouse | |
purified | |
RUO | |
3920, 695379 | |
4° C, store in dark | |
Liquid |