missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lass2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA112921
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control for this product is liver or kidney lysate.
Lass2 Polyclonal antibody specifically detects Lass2 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
Lass2 | |
Polyclonal | |
PBS with 0.08% sodium azide | |
Q924Z4, Q96G23 | |
CERS2 | |
Synthetic peptide corresponding to human Lass2 protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CERS2 | |
0610013I17Rik; AI225939; ceramide synthase 2; CERS2; FLJ10243; L3; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; LAG1 longevity assurance homolog 2; ceramide synthase 2; LASS2; longevity assurance (LAG1, S. cerevisiae) homolog 2; longevity assurance homolog 2; MGC987; SP260; Sphingosine N-acyltransferase CERS2; TMSG1; TRAM homolog 3; translocating chain-associating membrane protein homolog 3; Trh3; Tumor metastasis-suppressor gene 1 protein | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
29956, 310667, 76893 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |