Learn More
LC3B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA116930
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The reaction with mouse LC3 is weaker than human. The immunogen for this antibody production has 100% sequence homology with rat and zebrafish, 92% with mouse, and 91% with bovine proteins.
LC3B Polyclonal antibody specifically detects LC3B in Avian, Bacteria, Bovine, Canine, Chicken, Guinea Pig, Hamster, Human, Invertebrate, Mouse, Non-human Primate, Porcine, Rabbit, Rat, Zebrafish samples. It is validated for ChIP Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay (PLA), Western Blot.Spezifikation
LC3B | |
Polyclonal | |
Unconjugated | |
MAP1LC3B2 | |
1010001C15Rik; Atg8; ATG8E; ATG8F; ATG8G; Autophagy-related protein LC3 B; autophagy-related ubiquitin-like modifier LC3 B; LC3; LC3 beta; LC3A; LC3B; light chain 3; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 B; MAP1A/1BLC3; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; Map1alc3; MAP1BLC3; MAP1LC3; MAP1LC3A; MAP1LC3B; MAP1LC3B2; MAP1LC3B-a; microtubule associated protein 1 light chain 3 beta; microtubule associated protein 1 light chain 3 beta 2; microtubule-associated protein 1 light chain 3; microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3 beta 2; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3 beta 2; Microtubule-associated proteins 1A/1B light chain 3B; Microtubule-associated proteins 1A/1B light chain 3B-like; Microtubule-associated proteins 1A/1B light chain 3B-like protein; Mpl3; RP11-346K17.1; wu:fb60g11; zbs559; zgc:56434 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100349465, 100359928, 100462754, 100726328, 100769810, 322425, 408001, 427559, 479619, 643246, 64862, 67443, 81631 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
A6NCE7, O41515, Q62625, Q9CQV6, Q9GZQ8 | |
MAP1LC3B, MAP1LC3B2 | |
Synthetic peptide between amino acids 1-100 of the human LC3 protein. | |
100 μL | |
Primary | |
Avian, Non-human Primate, Porcine, Canine, Chicken, Guinea Pig, Hamster, Bovine, Invertebrate, Mouse, Guinea Pig, Hamster, Human, Invertebrate, Mouse | |
Antibody | |
IgG |