missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCMT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535913
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (88%), Rat (89%), Rhesus Monkey (98%), Bovine (90%).
LCMT1 Polyclonal antibody specifically detects LCMT1 in Human samples. It is validated for Immunocytochemistry.Spezifikation
LCMT1 | |
Polyclonal | |
Unconjugated | |
LCMT1 | |
[phosphatase 2A protein]-leucine-carboxy methyltransferase; [Phosphatase 2A protein]-leucine-carboxy methyltransferase 1; AL033290; CGI-68; Lcmt; Lcmt1; LCMT-1; Leucine carboxyl methyltransferase 1; PPMT1; protein phosphatase methyltransferase 1; protein-leucine O-methyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UIC8 | |
LCMT1 | |
Recombinant fragment corresponding to a region within amino acids 78 and 298 of LCMT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |