Learn More
LDB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521577
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa, HepG2, Mouse brain. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (97%), Pig (98%), Chicken (97%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LDB1 Polyclonal antibody specifically detects LDB1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
LDB1 | |
Polyclonal | |
Unconjugated | |
LDB1 | |
carboxy terminal LIM domain protein 2; carboxyl-terminal LIM domain-binding protein 2; CLIM2; CLIM-2; hLdb1; Ldb1; LDB-1; LIM domain binding 1; LIM domain-binding factor CLIM2; LIM domain-binding factor-1; LIM domain-binding protein 1; mLdb1; NLI; Nuclear LIM interactor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16825, 309447, 8861 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P70662, Q86U70 | |
LDB1 | |
Recombinant fragment corresponding to a region within amino acids 86 and 411 of LDB1 (Uniprot ID#Q86U70) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |