Learn More
LEF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA124462
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-24462 detects LEF1 in Human samples. PA1-24462 has been successfully used in Western Blot. PA1-24462 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human LEF1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Lymphoid enhancer-binding factor-1 (LEF1) is a 48-kD nuclear protein that is expressed in pre-B and T cells. It binds to a functionally important site in the T-cell receptor-alpha (TCRA) enhancer and confers maximal enhancer activity. LEF1 belongs to a family of regulatory proteins that share homology with high mobility group protein-1 (HMG1).Spezifikation
LEF1 | |
Polyclonal | |
Unconjugated | |
Q9UJU2 | |
LEF1 | |
Synthetic peptide region of human LEF1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
LEF1 | |
DKFZp586H0919, FLJ46390, TCF1ALPHA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51176 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |