missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LENG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546521
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: GDEGSRSRKE KEGSEKQRPK EPPSLDQLRA ERLRREAAER SRAEALLARV Sequence homology: Horse: 92%; Human: 100%
LENG1 Polyclonal antibody specifically detects LENG1 in Human samples. It is validated for Western Blot.Spezifikation
LENG1 | |
Polyclonal | |
Unconjugated | |
LENG1 | |
1500001K17Rik; Leng1; leukocyte receptor cluster (LRC) member 1; leukocyte receptor cluster member 1; Leukocyte receptor cluster member 1 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79165 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96BZ8 | |
LENG1 | |
Synthetic peptide directed towards the C-terminal of human LENG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |