Learn More
MBOAT7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543430
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: WWLAQYIYKS APARSYVLRS AWTMLLSAYW HGLHPGYYLS FLTIPLCLAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%
MBOAT7 Polyclonal antibody specifically detects MBOAT7 in Human samples. It is validated for Western Blot.Spezifikation
MBOAT7 | |
Polyclonal | |
Unconjugated | |
MBOAT7 | |
1-acylglycerophosphatidylinositol O-acyltransferase; 5730589L02Rik; BB1; Bladder and breast carcinoma-overexpressed gene 1 protein; h-mboa-7; hMBOA-7; Leng4; leukocyte receptor cluster (LRC) member 4; leukocyte receptor cluster member 4; LPIAT; Lpiat1; LPLAT 7; LRC4; lysophosphatidylinositol acyltransferase; Lysophosphatidylinositol acyltransferase 1; Lysophospholipid acyltransferase 7; lyso-PI acyltransferase; malignant cell expression-enhanced gene/tumor progression-enhanced; mBB1; MBOA7; Mboat7; membrane bound O-acyltransferase domain containing 7; membrane-bound O-acyltransferase domain-containing protein 7; m-mboa-7; OACT7; O-acyltransferase domain-containing protein 7; RGD1306945 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79143 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96N66 | |
MBOAT7 | |
Synthetic peptide directed towards the C-terminal of human LENG4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |