missing translation for 'onlineSavingsMsg'
Learn More

Leptin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA1051

466.45 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21653" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11521462

  • 491.00 € / 50 Mikroliter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1-051 detects leptin from human, mouse, sheep and rat samples. PA1-051 has been successfully used in Western blot, immunocytochemistry and immunohistochemistry experiments. By Western blot, this antibody detects an ∽16 kDa protein representing leptin from 3T3-L1 cell extract. Immunohistochemical staining of leptin in sheep brain with PA1-051 results in staining of the ventromedial hypothalamus. The PA1-051 immunogen is a synthetic peptide corresponding to residues Q(25) K V Q D D T K T L I K T I V T R I N D(44) of mouse Leptin. This sequence is completely conserved between mouse and bovine.

Leptin Polyclonal antibody specifically detects Leptin in Human, Mouse, Ovine, Rat samples. It is validated for ChIP Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Spezifikation

Spezifikation

Leptin
Polyclonal
Unconjugated
Lep
CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin
Rabbit
Antigen affinity chromatography
RUO
16846, 25608, 3952, 443534
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
2 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P41159, P41160, P50596, Q28603
Lep
Synthetic peptide corresponding to residues Q(25) K V Q D D T K T L I K T I V T R I N D(44) of mouse Leptin.
50 μL
Primary
Human, Mouse, Ovine, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote