Learn More
Leptin Monoclonal Antibody (022-11), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen ABS02211B01
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ABS 022-11B-01 detects Leptin in human samples. ABS 022-11B-01 has been successfully used in ELISA procedures. ABS 022-11B-01 binds free Leptin in solution. In sandwich ELISA, ABS 022-11B-01 works as a detection antibody with ABS 022-03-02 as capture antibody. The ABS 022-11B-01 immunogen is a purified, recombinant Leptin protein. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Leptin Monoclonal antibody specifically detects Leptin in Human samples. It is validated for ELISA.Spezifikation
Leptin | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P41159 | |
Lep | |
Purified, recombinant Leptin. plasma. | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
022-11 | |
Biotin | |
Lep | |
CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin | |
Mouse | |
purified | |
RUO | |
3952 | |
4° C, store in dark | |
Liquid |