Learn More
Leptin Monoclonal Antibody (11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen ABS0221102
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ABS 022-11-02 detects Leptin in human samples. ABS 022-11-02 has been successfully used in ELISA and Western blot procedures. ABS 022-11-02 binds free Leptin in solution. In sandwich ELISA, ABS 022-11-02 works as a detection antibody with ABS 022-03-02 as a capture antibody. ABS 022-11-02 shows three bands at 16, 32, and 48 kDa in the Western blot application corresponding with the leptin monomer, dimer and trimer forms. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Leptin Monoclonal antibody specifically detects Leptin in Human samples. It is validated for ELISA.Spezifikation
Leptin | |
Monoclonal | |
1.08 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P41159 | |
Lep | |
Purified, recombinant Leptin. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
11 | |
Unconjugated | |
Lep | |
CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin | |
Mouse | |
Protein A/G | |
RUO | |
3952 | |
4° C, store in dark | |
Liquid |