missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LFG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141589
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
LFG Polyclonal antibody specifically detects LFG in Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
LFG | |
Polyclonal | |
Unconjugated | |
FAIM2 | |
2900002L20Rik; FAIM2; Fas apoptotic inhibitory molecule 2; fas apoptotic inhibitory molecule 2; protein lifeguard 2; Kiaa0950; Lfg; LFG2; lifeguard; neural membrane protein 35; NGP35; NMP25; Nmp35; protein lifeguard; protein lifeguard 2; TMBIM2; transmembrane BAX inhibitor motif-containing protein 2 | |
Rabbit | |
Protein G | |
RUO | |
23017, 246274, 477613, 72393 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
O88407, Q8K097, Q9BWQ8 | |
FAIM2 | |
Synthetic peptide corresponding to residues 1-50 of human Lifeguard. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |