Learn More
LIN54 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531777
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (98%), Rat (97%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LIN54 Polyclonal antibody specifically detects LIN54 in Human samples. It is validated for Western Blot.Spezifikation
LIN54 | |
Polyclonal | |
Unconjugated | |
LIN54 | |
4832414L08; AI461788; CXC domain-containing protein 1; CXCDC1; JC8.6; KIAA2037; Lin54; lin-54 DREAM MuvB core complex component; lin-54 homolog; lin-54 homolog (C. elegans); MIP120; mKIAA2037; protein lin-54 homolog; RGD1311361; TCX1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.82 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6MZP7 | |
LIN54 | |
Recombinant fragment corresponding to a region within amino acids 377 and 621 of Human LIN54 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |