Learn More
LIN7A Monoclonal Antibody (P377), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA518298
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
Recommended positive controls: MCF-7, MDA-MB-231. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LIN7A Monoclonal antibody specifically detects LIN7A in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
LIN7A | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O14910 | |
LIN7A | |
Recombinant fragment corresponding to a region within amino acids 12 and 227 of LIN7A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
P377 | |
Unconjugated | |
LIN7A | |
AI848705; hLin-7; lin 7 homolog a; LIN7; lin-7 homolog A (C. elegans); lin-7 homolog A, crumbs cell polarity complex component; Lin7a; lin-7A; lin-7-Ba; Mals1; MALS-1; mammalian LIN-7 1; mammalian lin-seven protein 1; mLin-7; protein lin-7 homolog A; Tax interaction protein 33; TIP-33; Veli; Veli1; veli-1; veli-1 protein; vertebrate homolog of C. elegans Lin-7 type 1; vertebrate LIN7 homolog 1; vertebrate lin-7 homolog 1 | |
Mouse | |
Protein G | |
RUO | |
8825 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |