Learn More
LIPG Rabbit anti-Bovine, Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA132483
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-32483 detects Lipase from bovine, rat, mouse, human and Non-human primate samples. PA1-32483 has been successfully used in immunohistochemistry (paraffin tissue), Western blot applications. The PA1-32483 immunogen is: Synthetic peptide surrounding amino acid 22 of human Endothelial Lipase. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
LIPG (Lipase G, Endohelial type) is an endothelial lipase which is a novel member of the lipoprotein lipase gene family. LIPG is synthesized by endothelial cells and functions at the site of synthesis. LIPG most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed LIPG has been shown to reduce HDL cholesterol levels, whereas an LIPG deficiency appears to be effective in elevating HDL cholesterol levels in animals. LIPG has substantial phospholipase activity and may be involved in lipoprotein metabolism and vascular biology. Also, LIPG is designated a member of the TG lipase family by its sequence and characteristic lid region which provides substrate specificity for enzymes of the TG lipase family. LIPG is a carboxyl esterase that hydrolyzes high density lipoproteins, emulsifies triglycerides, is essential for the efficient digestion of dietary fats and can bind to heparin.Spezifikation
LIPG | |
Polyclonal | |
Unconjugated | |
Q9Y5X9, Q9WVG5, Q8VBX1 | |
LIPG | |
Synthetic peptide surrounding amino acid 22 of human Endothelial Lipase. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
LIPG | |
lipase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16891, 291437, 509808, 9388 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |