Learn More
Lipoteichoic Acid Monoclonal Antibody (55), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA140134
Weitere Details : Netto-Gewicht : 1.24740kg
Beschreibung
MA1-40134 detects Lipoteichic Acid from gram positive bacterial samples. MA1-40134 has been successfully used in Western blot, ELISA, flow cytometry, immunofluorescence, and Immunohistochemistry (frozen) applications. Positive control: Culture medium of Gram-positive bacteria. Negative control: Culture medium of eukaryotic cells. The MA1-40134 immunogen is a microbial mixture of Streptococcus sobrinus HG961, HG962, HG970, and HG977.
Lipoteichoic Acid Monoclonal antibody specifically detects Lipoteichoic Acid in Bacteria samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
Lipoteichoic Acid | |
Monoclonal | |
>200 μg/mL | |
tissue culture supernatant with 0.02% sodium azide | |
Mouse | |
1 mL | |
Primary | |
4° C | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
55 | |
Unconjugated | |
LTA | |
Microbial mixture of Streptococcus sobrinus HG961, HG962, HG970, and HG977 | |
RUO | |
Bacteria | |
Antibody | |
IgG3 |