missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA126408
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-26408 detects liver Arginase from bovine samples.
Arginase 1 Polyclonal antibody specifically detects Arginase 1 in Bovine samples. It is validated for Immunomicroscopy, Western Blot.Spezifikation
Arginase 1 | |
Polyclonal | |
Unconjugated | |
ARG1 | |
AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase | |
Rabbit | |
IgG fraction | |
RUO | |
513608 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunomicroscopy, Western Blot | |
10 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2 | |
Q2KJ64 | |
ARG1 | |
Arginase from bovine liver. | |
1 mg | |
Primary | |
Bovine | |
Antibody | |
IgG |