missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CES1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519740
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with baboon and rhesus monkey based on sequence homology. Store antibody at 4oC for 1-2 weeks. For long-term storage, store at -20oC.
CES1 Polyclonal antibody specifically detects CES1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
CES1 | |
Polyclonal | |
Unconjugated | |
CES1 | |
ACAT; acyl coenzyme A:cholesterol acyltransferase; acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; carboxyesterase ES-3; carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1A; carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 1-like; carboxylesterase 2 (liver); CE-1; CEH; Ces1; Ces-1; Ces1a; Ces1e; Ces1g; Ces1l; CES2; cholesteryl ester hydrolase; cocaine carboxylesterase; Eg; EG244595; Egasyn; Es22; Es-22; ES-HTEL; esterase 22; esterase-22; ES-x; Gm4976; hCE-1; HMSE; HMSE1; human monocyte/macrophage serine esterase 1; liver carboxylesterase 1; Liver carboxylesterase 22; liver carboxylesterase 3; methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; PCE-1; pI 5.5 esterase; REH; Retinyl ester hydrolase; serine esterase 1; SES1; Ses-1; TGH; triacylglycero; Triacylglycerol hydrolase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1066, 12623, 13884, 13897, 24346, 244595, 29225, 679817 | |
-20°C or -80°C if preferred | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
P10959, P23141, P23953, Q63108, Q64176, Q8VCC2 | |
CES1, Ces1a, Ces1c, Ces1e, Ces1g | |
Synthetic peptide conjugated to KLH derived from within residues 500 to the C-terminus of Human Liver Carboxylesterase 1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |