missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LKB1 Monoclonal Antibody (E.425.6), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA514831
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
LKB1 Monoclonal antibody specifically detects LKB1 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.Spezifikation
LKB1 | |
Monoclonal | |
24 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
D4AE59, Q15831, Q9WTK7 | |
STK11 | |
An LKB1 partial fusion protein | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Western Blot | |
E.425.6 | |
Unconjugated | |
STK11 | |
AA408040; CAMK group; CAMKL family; hLKB1; liver kinase B1; Liver kinase B1 homolog; LKB 1; LKB1; LKB-1; LKB1(S); Par-4; PJS; polarization-related protein LKB1; R75140; renal carcinoma antigen NY-REN-19; serine/threonine kinase 11; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; serine/threonine-protein kinase STK11; STK11 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20869, 314621, 6794 | |
-20°C | |
Liquid |