Learn More
LMAN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530081
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2, HCT116. Predicted reactivity: Mouse (93%), Rat (93%), Chicken (81%), Rhesus Monkey (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LMAN1 Polyclonal antibody specifically detects LMAN1 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
LMAN1 | |
Polyclonal | |
Unconjugated | |
LMAN1 | |
2610020P13Rik; AI326273; AU043785; C730041J05; EGK_09725; endoplasmic reticulum-golgi intermediate compartment protein 53; ERGIC53; ERGIC-53; ER-Golgi intermediate compartment 53 kDa protein; F5F8D; FMFD1; Gp58; intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose binding 1; lectin, mannose-binding, 1; LMAN1; MCFD1; MR60; P58; Protein ERGIC-53 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3998 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.82 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P49257 | |
LMAN1 | |
Recombinant fragment corresponding to a region within amino acids 21 and 282 of Human LMAN1 | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |